Recombinant Mouse TGFB1 (from E. coli), His-tag
Catalog # ABNOP7768.250UG
Supplier: Abnova
CAS Number:
Specifications
- VE:250 µG
- Protein/Peptid-Typ:Rekombinant
- Ursprung:E. coli
- Spezies:Mouse
- Tag sequence:His-tag
- Gen-ID:NP_035707
- Protein/Peptid-Name:TGFB1
- Sequence:279a.a.-390a.a