Recombinant Human COMP (from HEK293 cells), His-tag
Catalog # ABNOP7867.500UG
Supplier: Abnova
CAS Number:
Specifications
- VE:500 µG
- Protein/Peptid-Typ:Rekombinant
- Ursprung:HEK293 cells
- Spezies:Human
- Tag sequence:His-tag
- Gen-ID:P49747
- Protein/Peptid-Name:COMP
- Sequence:21a.a.-757a.a.