Order Entry
Austria
ContactUsLinkComponent
 
Recombinant Human MRPL2 (from E. coli), His-tag
  ABNOP7754.500UG
 :  Abnova
 :  
Recombinant Human MRPL2 (from E. coli), His-tag
  ABNOP7754.500UG
 :  Abnova
 :  

 

  • VE:
    500 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Human
  • Tag sequence:
    His-tag
  • Gen-ID:
    NP_0570341
  • Protein/Peptid-Name:
    MRPL2
  • Sequence:
    82a.a.-202a.a