ContactUsLinkComponent
Recombinant Rat IL10 (from E. coli)
Recombinant Rat IL10 (from E. coli)
Catalog # ABNOP7627.1MG
Supplier:  Abnova
CAS Number:  
Recombinant Rat IL10 (from E. coli)
Catalog # ABNOP7627.1MG
Supplier:  Abnova
CAS Number:  
Specifications
  • VE:
    1 mg
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Rat
  • Gen-ID:
    P29456
  • Protein/Peptid-Name:
    IL10
  • Sequence:
    19a.a.-178a.a.