Order Entry
Austria
ContactUsLinkComponent
Recombinant Mouse CNTF (from E. coli)
Recombinant Mouse CNTF (from E. coli)
  ABNOP7642.10UG
 :  Abnova
 :  
Recombinant Mouse CNTF (from E. coli)
  ABNOP7642.10UG
 :  Abnova
 :  

 

  • VE:
    10 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Mouse
  • Gen-ID:
    P51642
  • Protein/Peptid-Name:
    CNTF
  • Sequence:
    2a.a.-198a.a.