Order Entry
Austria
ContactUsLinkComponent
Recombinant Mouse IL1A (from E. coli), His-tag
Recombinant Mouse IL1A (from E. coli), His-tag
Catalog # ABNOP7713.500UG
Supplier:  Abnova
CAS Number:  
Recombinant Mouse IL1A (from E. coli), His-tag
Catalog # ABNOP7713.500UG
Supplier:  Abnova
CAS Number:  

Specifications

  • VE:
    500 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Mouse
  • Tag sequence:
    His-tag
  • Gen-ID:
    NP_034684
  • Protein/Peptid-Name:
    IL1A
  • Sequence:
    115a.a.-270a.a

Specifications

About this item