Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteine und Peptide
Recombinant Mouse IL9
Recombinant Mouse IL9
ABNOP7561.10UG
:
Abnova
:
Recombinant Mouse IL9
ABNOP7561.10UG
:
Abnova
:
VE:
10 µG
Protein/Peptid-Typ:
Rekombinant
Spezies:
Mouse
Gen-ID:
P15247
Protein/Peptid-Name:
IL9
Sequence:
19a.a.-144a.a.