Recombinant Human CTLA4 (from CHO cells), hFc tag
Catalog # ABNOP7539.1MG
Supplier: Abnova
CAS Number:
Specifications
- VE:1 mg
- Protein/Peptid-Typ:Rekombinant
- Ursprung:CHO cells
- Spezies:Human
- Tag sequence:hFc tag
- Gen-ID:P16410
- Protein/Peptid-Name:CTLA4
- Sequence:37a.a.-162a.a.