Recombinant Human CD38 (from HEK293 cells)
Catalog # ABNOP7553.1MG
Supplier: Abnova
CAS Number:
Specifications
- VE:1 mg
- Protein/Peptid-Typ:Rekombinant
- Ursprung:HEK293 cells
- Spezies:Human
- Gen-ID:P28907
- Protein/Peptid-Name:CD38
- Sequence:43a.a.-300a.a.