Recombinant Human TGFB1 (from CHO cells)
Catalog # ABNOP7562.10UG
Supplier: Abnova
CAS Number:
Specifications
- VE:10 µG
- Protein/Peptid-Typ:Rekombinant
- Ursprung:CHO cells
- Spezies:Human
- Gen-ID:P01137
- Protein/Peptid-Name:TGFB1
- Sequence:279a.a.-390a.a.