Recombinant Mouse IL22 (from E. coli)
Catalog # ABNOP7509.10UG
Supplier: Abnova
CAS Number:
Specifications
- VE:10 µG
- Protein/Peptid-Typ:Rekombinant
- Ursprung:E. coli
- Spezies:Mouse
- Gen-ID:Q9JJY9
- Protein/Peptid-Name:IL22
- Sequence:34a.a.-179a.a.