Order Entry
Austria
ContactUsLinkComponent
 
Recombinant Human TGFA (from E. coli)
  ABNOP7519.10UG
 :  Abnova
 :  
Recombinant Human TGFA (from E. coli)
  ABNOP7519.10UG
 :  Abnova
 :  

 

  • VE:
    10 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Human
  • Protein/Peptid-Name:
    TGFA
  • Sequence:
    HumanTGFA(40a.a.-89a.a.