Order Entry
Austria
ContactUsLinkComponent
Recombinant Mouse KLK7 (from CHO cells), His-tag
Recombinant Mouse KLK7 (from CHO cells), His-tag
Catalog # ABNOP7608.50UG
Supplier:  Abnova
CAS Number:  
Recombinant Mouse KLK7 (from CHO cells), His-tag
Catalog # ABNOP7608.50UG
Supplier:  Abnova
CAS Number:  

Specifications

  • VE:
    50 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    CHO cells
  • Spezies:
    Mouse
  • Tag sequence:
    His-tag
  • Gen-ID:
    Q91VE3
  • Protein/Peptid-Name:
    KLK7
  • Sequence:
    22a.a.-249a.a.

Specifications

About this item