Order Entry
Austria
ContactUsLinkComponent
Recombinant Human CD276 (from HEK293 cells), hFc tag
Recombinant Human CD276 (from HEK293 cells), hFc tag
Catalog # ABNOP7576.1MG
Supplier:  Abnova
CAS Number:  
Recombinant Human CD276 (from HEK293 cells), hFc tag
Catalog # ABNOP7576.1MG
Supplier:  Abnova
CAS Number:  

Specifications

  • VE:
    1 mg
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    HEK293 cells
  • Spezies:
    Human
  • Tag sequence:
    hFc tag
  • Gen-ID:
    Q5ZPR3-2
  • Protein/Peptid-Name:
    CD276
  • Sequence:
    29a.a.-245a.a.

Specifications

About this item