Order Entry
Austria
ContactUsLinkComponent
Anti-RELA Rabbit Recombinant Antibody [clone: NFKBP65S468-B9]
Anti-RELA Rabbit Recombinant Antibody [clone: NFKBP65S468-B9]
Catalog # ABNORAB02864.200UL
Supplier:  Abnova
Anti-RELA Rabbit Recombinant Antibody [clone: NFKBP65S468-B9]
Catalog # ABNORAB02864.200UL
Supplier:  Abnova

Specifications

  • Antigen-Symbol:
    RELA
  • Klonalität:
    Recombinant
  • Klon:
    NFKBP65S468-B9
  • Wirt:
    Rabbit
  • Reaktivität:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human RELA.
  • VE:
    200 µl

Specifications

About this item