Anti-Phl p 1 Human Recombinant Antibody [clone: 25]
Catalog # ABNORAB03756.50UG
Supplier: Abnova
Specifications
- Antigen-Symbol:Phl p 1
- Klonalität:Recombinant
- Klon:25
- Wirt:Human
- Reaktivität:Timothy grass (Phleum pratense)
- Immunogen:Recombinant human monoclonal antibody raised against Timothy-Grass Phl p 1.
- VE:50 µG