Order Entry
Austria
ContactUsLinkComponent
Anti-POU2F1 Rabbit Recombinant Antibody [clone: R03-1K1]
Catalog # ABNORAB02041.100UL
Supplier:  Abnova
Anti-POU2F1 Rabbit Recombinant Antibody [clone: R03-1K1]
Catalog # ABNORAB02041.100UL
Supplier:  Abnova

Specifications

  • Antigen-Symbol:
    POU2F1
  • Klonalität:
    Recombinant
  • Klon:
    R03-1K1
  • Wirt:
    Rabbit
  • Reaktivität:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human POU2F1.
  • VE:
    100 µl

Specifications

About this item