Anti-RAB5A Rabbit Recombinant Antibody [clone: R06-4J3]
Catalog # ABNORAB01967.100UL
Supplier: Abnova
Specifications
- Antigen-Symbol:RAB5A
- Klonalität:Recombinant
- Klon:R06-4J3
- Wirt:Rabbit
- Reaktivität:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human RAB5A.
- VE:100 µl