Order Entry
Austria
ContactUsLinkComponent
Anti-PFKFB3 Rabbit Recombinant Antibody [clone: R08-7A4]
Anti-PFKFB3 Rabbit Recombinant Antibody [clone: R08-7A4]
Catalog # ABNORAB02009.100UL
Supplier:  Abnova
Anti-PFKFB3 Rabbit Recombinant Antibody [clone: R08-7A4]
Catalog # ABNORAB02009.100UL
Supplier:  Abnova

Specifications

  • Antigen-Symbol:
    PFKFB3
  • Klonalität:
    Recombinant
  • Klon:
    R08-7A4
  • Wirt:
    Rabbit
  • Reaktivität:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human PFKFB3.
  • VE:
    100 µl

Specifications

About this item