Order Entry
Austria
ContactUsLinkComponent
 
Anti-PDK2 Rabbit Recombinant Antibody [clone: R01-6D8]
Catalog # ABNORAB02014.100UL
Supplier:  Abnova
Anti-PDK2 Rabbit Recombinant Antibody [clone: R01-6D8]
Catalog # ABNORAB02014.100UL
Supplier:  Abnova

Specifications

  • Antigen-Symbol:
    PDK2
  • Klonalität:
    Recombinant
  • Klon:
    R01-6D8
  • Wirt:
    Rabbit
  • Reaktivität:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human PDK2.
  • VE:
    100 µl

Specifications

About this item