Order Entry
Austria
ContactUsLinkComponent
Anti-PCSK9 Rabbit Recombinant Antibody [clone: R09-4F7]
Catalog # ABNORAB02017.100UL
Supplier:  Abnova
Anti-PCSK9 Rabbit Recombinant Antibody [clone: R09-4F7]
Catalog # ABNORAB02017.100UL
Supplier:  Abnova

Specifications

  • Antigen-Symbol:
    PCSK9
  • Klonalität:
    Recombinant
  • Klon:
    R09-4F7
  • Wirt:
    Rabbit
  • Reaktivität:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human PCSK9.
  • VE:
    100 µl

Specifications

About this item