Anti-PARP1 Rabbit Recombinant Antibody [clone: R05-8E8]
Catalog # ABNORAB02022.100UL
Supplier: Abnova
Specifications
- Antigen-Symbol:PARP1
- Klonalität:Recombinant
- Klon:R05-8E8
- Wirt:Rabbit
- Reaktivität:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human PARP1.
- VE:100 µl