Anti-PRKCH Rabbit Recombinant Antibody [clone: R03-7A9]
ABNORAB01993.100UL
: Abnova
- Antigen-Symbol:PRKCH
- Klonalität:Recombinant
- Klon:R03-7A9
- Wirt:Rabbit
- Reaktivität:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human PRKCH.
- VE:100 µl