Anti-HBEGF Rabbit Recombinant Antibody [clone: R08-9C2]
Catalog # ABNORAB02233.100UL
Supplier: Abnova
Specifications
- Antigen-Symbol:HBEGF
- Klonalität:Recombinant
- Klon:R08-9C2
- Wirt:Rabbit
- Reaktivität:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human HBEGF.
- VE:100 µl