Anti-GLS Rabbit Recombinant Antibody [clone: R08-3D8]
ABNORAB02251.100UL
: Abnova
- Antigen-Symbol:GLS
- Klonalität:Recombinant
- Klon:R08-3D8
- Wirt:Rabbit
- Reaktivität:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human GLS.
- VE:100 µl