Order Entry
Austria
ContactUsLinkComponent
Anti-GFPT1 Rabbit Recombinant Antibody [clone: R01-2I2]
Anti-GFPT1 Rabbit Recombinant Antibody [clone: R01-2I2]
Catalog # ABNORAB02256.100UL
Supplier:  Abnova
Anti-GFPT1 Rabbit Recombinant Antibody [clone: R01-2I2]
Catalog # ABNORAB02256.100UL
Supplier:  Abnova

Specifications

  • Antigen-Symbol:
    GFPT1
  • Klonalität:
    Recombinant
  • Klon:
    R01-2I2
  • Wirt:
    Rabbit
  • Reaktivität:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human GFPT1.
  • VE:
    100 µl

Specifications

About this item