Order Entry
Austria
ContactUsLinkComponent
Anti-GDF15 Rabbit Recombinant Antibody [clone: R04-8I1]
Catalog # ABNORAB02260.100UL
Supplier:  Abnova
Anti-GDF15 Rabbit Recombinant Antibody [clone: R04-8I1]
Catalog # ABNORAB02260.100UL
Supplier:  Abnova

Specifications

  • Antigen-Symbol:
    GDF15
  • Klonalität:
    Recombinant
  • Klon:
    R04-8I1
  • Wirt:
    Rabbit
  • Reaktivität:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human GDF15.
  • VE:
    100 µl

Specifications

About this item