Order Entry
Austria
ContactUsLinkComponent
Anti-TFEB Rabbit Recombinant Antibody [clone: R05-0D7]
Anti-TFEB Rabbit Recombinant Antibody [clone: R05-0D7]
Catalog # ABNORAB01345.100UL
Supplier:  Abnova
Anti-TFEB Rabbit Recombinant Antibody [clone: R05-0D7]
Catalog # ABNORAB01345.100UL
Supplier:  Abnova

Specifications

  • Antigen-Symbol:
    TFEB
  • Klonalität:
    Recombinant
  • Klon:
    R05-0D7
  • Wirt:
    Rabbit
  • Reaktivität:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human TFEB.
  • VE:
    100 µl

Specifications

About this item