Anti-KAT7 Rabbit Recombinant Antibody [clone: R01-2I6]
ABNORAB01423.100UL
: Abnova
- Antigen-Symbol:KAT7
- Klonalität:Recombinant
- Klon:R01-2I6
- Wirt:Rabbit
- Reaktivität:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human KAT7.
- VE:100 µl