Anti-HMGB2 Rabbit Recombinant Antibody [clone: R02-5I5]
ABNORAB01430.100UL
: Abnova
- Antigen-Symbol:HMGB2
- Klonalität:Recombinant
- Klon:R02-5I5
- Wirt:Rabbit
- Reaktivität:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human HMGB2.
- VE:100 µl