Order Entry
Austria
ContactUsLinkComponent
Anti-GRM2 Rabbit Recombinant Antibody [clone: R04-2D1]
Catalog # ABNORAB02106.100UL
Supplier:  Abnova
Anti-GRM2 Rabbit Recombinant Antibody [clone: R04-2D1]
Catalog # ABNORAB02106.100UL
Supplier:  Abnova

Specifications

  • Antigen-Symbol:
    GRM2
  • Klonalität:
    Recombinant
  • Klon:
    R04-2D1
  • Wirt:
    Rabbit
  • Reaktivität:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human GRM2.
  • VE:
    100 µl

Specifications

About this item