Order Entry
Austria
ContactUsLinkComponent
Anti-SYNE1 Rabbit Recombinant Antibody [clone: R07-4E7]
  ABNORAB02069.100UL
 :  Abnova
Anti-SYNE1 Rabbit Recombinant Antibody [clone: R07-4E7]
  ABNORAB02069.100UL
 :  Abnova

 

  • Antigen-Symbol:
    SYNE1
  • Klonalität:
    Recombinant
  • Klon:
    R07-4E7
  • Wirt:
    Rabbit
  • Reaktivität:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human SYNE1.
  • VE:
    100 µl