Anti-NEK7 Rabbit Recombinant Antibody [clone: R04-7A1]
Catalog # ABNORAB02071.100UL
Supplier: Abnova
Specifications
- Antigen-Symbol:NEK7
- Klonalität:Recombinant
- Klon:R04-7A1
- Wirt:Rabbit
- Reaktivität:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human NEK7.
- VE:100 µl