Anti-TRIM28 Rabbit Recombinant Antibody [clone: R07-5E6]
Catalog # ABNORAB02152.100UL
Supplier: Abnova
Specifications
- Antigen-Symbol:TRIM28
- Klonalität:Recombinant
- Klon:R07-5E6
- Wirt:Rabbit
- Reaktivität:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human TRIM28.
- VE:100 µl