Order Entry
Austria
ContactUsLinkComponent
Anti-MAPK9 Rabbit Recombinant Antibody [clone: R02-9H4]
  ABNORAB02155.100UL
 :  Abnova
Anti-MAPK9 Rabbit Recombinant Antibody [clone: R02-9H4]
  ABNORAB02155.100UL
 :  Abnova

 

  • Antigen-Symbol:
    MAPK9
  • Klonalität:
    Recombinant
  • Klon:
    R02-9H4
  • Wirt:
    Rabbit
  • Reaktivität:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human MAPK9.
  • VE:
    100 µl