Ultrapure water systems, PURELAB® Option Q
Catalog # ELGAOQ015BPM1
Supplier: ELGA LabWater
Specifications
- Fließrate:0,25 l/min
- Bezeichnung:PURELAB Option Q 15, with boost pump, flow rate 15 l/hr
- Seq:04
- Modell:Option Q-15
- Bakterien:<1 CFU/ml
- Leitfähigkeit:<1400 µS/cm
- Speisewasser:Potable mains water supply
- Spezifischer Widerstand bei +25 °C:up to 18,2 MΏ/cm
- TOC-Gehalt:1 - 3 ppb
- Gewicht:21 kg
- B×T×H:550×270×460 mm
- VE:1
Specifications
About this item
The PURELAB Option Q has been developed by ELGA to produce ultrapure water (type I) without the need to pre-purify the supply. The system can produce up to 20 L/hour (160 L/day) of water at 18,2 MΩcm, directly from a source of drinking water. Recirculating purified water within the DV25 docking vessel or reservoir ensures that the water purity is constantly at its maximum level.
- Microprocessor-controlled system management with continuous water purity monitoring
- Adjustable audio-visual alarms
- Direct access to front inspection doors for easier access to parts
- Cartridge replacement indicator
- RS232 interface enables data collection (GLP-compliant)
- ELGA Biofilter (optional) ensures that the water produced is free from endotoxins, bacteria and nucleases
Ordering information: Available with or without internal boost pump.