Relay Module
Relay module potential-free forwarding of 4 different alarm states
Catalog # MSGA38766
Supplier: asecos
Jump to:
Product Details & Documents
By choosing this option, you are opting for alarm differentiation instead of a collective alarm.
These signals are passed on to a central control centre via potential-free alarm contacts. With the clear differentiation of the alarm conditions, you can initiate optimally coordinated measures.
The relay module is suited for the PRO models of the IONLINE.
Specifications
- Bezeichnung:Relay module potential-free forwarding of 4 different alarm states
- Seq:166