Corning® Gosselin™ Petri Dish, Aseptic
Catalog # GOSSSH91-101
Supplier: Corning
Specifications
- Bezeichnung:Petri Dish 100×15 mm
- Environmentally Preferable:Umweltschonende Herstellung
- Umweltzertifizierung:RECs, ISO 14001 - Borre
- VE:700
Specifications
About this item
Transparent PS Petri dishes for food industry laboratories.
- Diam. 90 mm
- Crystal Polystyrene (PS)
- Optimal flatness
- Easy opening sleeves
- 1 vent
- Aseptic