Clax Magic Multi 70C2 Stain Remover
Catalog # DIVY100883039
Supplier: Diversey
Specifications
- Bezeichnung:Spot remover
- Verpackt:6×0,75 L
- VE:6 Bottle
- pH-Wert:8,2
Specifications
About this item
Clax Magic Multi is a stain remover with excellent properties to remove a variety of stains such as make-up, lipstick, ink and curry stains. The product is specially formulated for use in commercial and on premise laundries and is ready to use on most types of fabric.
- Blend of solvents and other cleaning components
- Consists of four innovative stain-fighting solutions