0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Gas-Spürsysteme
Accessories for Gas Detection System
Accessories for Gas Detection System
Catalog # DRAE6811535
Supplier:
Dräger
Expand image
Accessories for Gas Detection System
Catalog # DRAE6811535
Supplier:
Dräger
Specifications
Messbereich:
0 - 200 ppm
Bezeichnung:
XXS OV-A sensor, for use with the X-am 5000/5600 instruments
VE:
1
Specifications
About this item
Product Details
Product Details