Maintenaince Kit for Star Desiccators
BOHLV1778-02
- Bezeichnung:Maintenaince kit
- VE:1 KIT
- Für:SICCO Star desiccators
Maintenance Kit for SICCO Star-Desiccators consists of one cellular rubber sealing tape, one magnetic tape and four non-slip rubber feet.
- Easy replacement of sealing and magnetic tape to ensure the airtight closure of the desiccator