Scotch-Weld™ DP804 EPX Super Clear Acrylic Adhesive
Catalog # MMMADP804
Supplier: 3M
Specifications
- Volumen:48,5 ml
- Farbe:Transparent
- Bezeichnung:Scotch-Weld™ EPX Super clear acrylic adhesive DP804
- VE:12
Specifications
About this item
The Scotch-Weld™ EPX™ Bonding system is a discharging system for mixing two-component mixed adhesives. The main agent and the curing agent with the cartridge are mixed and discharged by a special hand gun and a special nozzle.
- Easy mixing and application with the help of the EPX gun
- 3 minutes of working time/4 minutes of handling time
- Exposure at temperatures from 0 to 100 °C
- Easy to apply
Stable adhesion can be obtained by eliminating the hassle of troublesome six-liquid mixing.