Oil Sorbent Boom
Catalog # MMMAT270GA
Supplier: 3M
Specifications
- Bezeichnung:Oil sorbent boom, T-270GA
- Länge:5 m
- VE:2
- Ø ext.:200 mm
Specifications
About this item
3M™ Oil sorbents are an innovative range of highly absorbent, hydrophobic (water repellent), quality products.
- Ideal for absorbing oil from areas where water may also be present
- Ideal for placing around equipment
- Prevent leaks or spills from spreading to adjacent areas
Will not disintegrate or sink, even when saturated with oil, and very easy to dispose of.