Culti-Loops™ Peptostreptococcus anaerobius ATCC™ 27337™
OXOIR4607051
:
- VE:5
- Bezeichnung:Peptostreptococcus anaerobius ATCC® 27337
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium.
: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.