Culti-Loops™ Paenibacillus polymyxa ATCC™ 43865™
OXOIR4601234
:
- VE:5
- Bezeichnung:Paenibacillus polymyxa ATCC® 43865™
- Seq:44
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.