Culti-Loops™ Campylobacter jejuni subsp. Jejuni ATCC™ 29428™
Artikelnr. OXOIR4601410
Lieferant: THERMO FISHER DIAGNOSTICS
CAS-Nummer:
Spezifikationen
- VE:5
- Bezeichnung:Campylobacter jejuni subsp. Jejuni ATCC™ 29428™
- Seq:62
Spezifikationen
Über diesen Artikel
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Packaging: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.