Order Entry
Austria
ContactUsLinkComponent
Anti-NFKBIA Tyr305 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-5515R-A750
Supplier:  Bioss
Anti-NFKBIA Tyr305 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-5515R-A750
Supplier:  Bioss

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    NFKBIA Tyr305
  • Klonalität:
    Polyklonal
  • Konjugation:
    Alexa Fluor® 750
  • Durchflusszytometrie:
    Yes
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • ImmunFluoreszenz:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gen-ID:
    4792
  • Antigen-Synonyme:
    MAD3|IkappaBalpha|IkB-alpha|Major histocompatibility complex enhancer-binding protein MAD3|NFKBI|NF-kappa-B inhibitor alpha|NFKBIA|I-kappa-B-alpha|MAD-3|IKBA
  • Modification:
    Phosphorylation (Tyr305)
  • Lagerungspuffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    35 kDa
  • Lagertemperatur:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Konzentration:
    1 µg/µl
  • Transporttemperatur:
    4 °C
  • Immunogen:
    KLH conjugated synthetic phosphopeptide derived from human IKB alpha around the phosphorylation site of Tyr305
  • Validierte Anwendungen:
    ICC
  • Reinigung:
    Purified by Protein A
  • Packung:
    Vial
  • VE:
    100 µl

Specifications

About this item

Inhibits the activity of dimeric NF-kappa-B/REL complexes by trapping REL dimers in the cytoplasm through masking of their nuclear localisation signals. On cellular stimulation by immune and proinflammatory responses, becomes phosphorylated promoting ubiquitination and degradation, enabling the dimeric RELA to translocate to the nucleus and activate transcription.

Type: Primary
Antigen: NFKBIA Tyr305
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat