Order Entry
Austria
ContactUsLinkComponent
Anti-CD74 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-CD74 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-2518R-A750
Supplier:  Bioss
Anti-CD74 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-2518R-A750
Supplier:  Bioss

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    CD74
  • Klonalität:
    Polyklonal
  • Konjugation:
    Alexa Fluor® 750
  • Durchflusszytometrie:
    Yes
  • Wirt:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    151-250/296
  • Form:
    Liquid
  • Gen-ID:
    972
  • Antigen-Synonyme:
    HLADG|II|p33|HLA class II histocompatibility antigen gamma chain|CD74|Ia antigen-associated invariant chain|DHLAG|Ia-GAMMA|HLA-DR antigens-associated invariant chain
  • Modification:
    Unmodified
  • Lagerungspuffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    34 kDa
  • Lagertemperatur:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Konzentration:
    1 µg/µl
  • Transporttemperatur:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human CD74
  • Reinigung:
    Purified by Protein A
  • Packung:
    Vial
  • VE:
    100 µl

Specifications

About this item

Plays a critical role in MHC class II antigen processing by stabilizing peptide-free class II alpha/beta heterodimers in a complex soon after their synthesis and directing transport of the complex from the endoplasmic reticulum to the endosomal/lysosomal system where the antigen processing and binding of antigenic peptides to MHC class II takes place. Serves as cell surface receptor for the cytokine MIF.

Type: Primary
Antigen: CD74
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 151-250/296
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse