- Antikörper-Typ:Primär
- Antigen-Name:IKK beta Tyr199
- Klonalität:Polyklonal
- Konjugation:Alexa Fluor® 750
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:3551
- Antigen-Synonyme:IKK-beta|I-kappa-B kinase 2|IMD15|IKK-B|IkBKB|IKKB|Inhibitor of nuclear factor kappa-B kinase subunit beta|I-kappa-B-kinase beta|Nuclear factor NF-kappa-B inhibitor kinase beta|IKK2|NFKBIKB
- Modification:Phosphorylation (Tyr199)
- Lagerungspuffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:87 kDa
- Lagertemperatur:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Konzentration:1 µg/µl
- Transporttemperatur:4 °C
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human IKK beta around the phosphorylation site of Tyr199
- Validierte Anwendungen:ICC
- Reinigung:Purified by Protein A
- Packung:Vial
- VE:100 µl
Serine kinase that plays an essential role in the NF-kappa-B signaling pathway which is activated by multiple stimuli such as inflammatory cytokines, bacterial or viral products, DNA damages or other cellular stresses. Acts as part of the canonical IKK complex in the conventional pathway of NF-kappa-B activation and phosphorylates inhibitors of NF-kappa-B on 2 critical serine residues. These modifications allow polyubiquitination of the inhibitors and subsequent degradation by the proteasome. In turn, free NF-kappa-B is translocated into the nucleus and activates the transcription of hundreds of genes involved in immune response, growth control, or protection against apoptosis. In addition to the NF-kappa-B inhibitors, phosphorylates several other components of the signaling pathway including NEMO/IKBKG, NF-kappa-B subunits RELA and NFKB1, as well as IKK-related kinases TBK1 and IKBKE. IKK-related kinase phosphorylations may prevent the overproduction of inflammatory mediators since they exert a negative regulation on canonical IKKs. Also phosphorylates other substrates including NCOA3, BCL10 and IRS1. Within the nucleus, acts as an adapter protein for NFKBIA degradation in UV-induced NF-kappa-B activation.
Type: Primary
Antigen: IKK beta Tyr199
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat