FailSafe™ PCR Enzyme and 2X Premix Buffers, PCR Optimisation Components
Catalog # EPICFSE51100
Supplier: EPICENTRE
CAS Number:
Specifications
- VE:1
- Bezeichnung:FailSafe™ Enzyme mix only
- Seq:02
- Größe:100 units
Specifications
About this item
Purchase individual FailSafe™ DNA polymerase and/or FailSafe™ 2X PreMix Buffers here.
- Trusted: The same great performance as the FailSafe™ PCR System, but available as individual components
- Flexible: Purchase any combination of FailSafe™ enzyme and/or 2X PreMixes you desire
This listing is only for reordering of individual FailSafe™ PCR 2X PreMixes or the standalone FailSafe™ DNA polymerase.
Before purchasing these components, we strongly recommend first optimising your PCR reaction conditions using the complete FailSafe™ PCR System in order to know which components to purchase based on their performance.
See the FailSafe™ PCR Systems webpage for more details on this system and PCR components.
Certifications: For research use only. Not for human or diagnostic use.
Ordering information: Each component listed here can be ordered separately and at any quantity. Each 2X Premix includes dNTPs, buffer, and various amounts of MgCl₂ and FailSafe™ PCR Enhancer with Betaine.